Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Xanthotoxin
Synonym(s):
8-Methoxypsoralen, 8-MOP, 9-Methoxyfuro[3,2-
g
][1]benzopyran-7-one, Ammoidin, Methoxsalen, Xanthotoxin
Empirical Formula (Hill Notation):
C
12
H
8
O
4
CAS Number:
298-81-7
Molecular Weight:
216.19
EC Number:
206-066-9
Products
Brand
Supelco (1)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
analytical standard (2)
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
PHL89866
Xanthotoxin,
phyproof
®
Reference Substance
Expand
View Pricing
56448
Xanthotoxin,
analytical standard
Expand
View Pricing