Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Whatman
®
glass microfiber filters, Grade 934-AH
®
RTU
Products
Product Category
filters (15)
Manufacturer
Diameter (mm)
Pore size (µm)
Filter Products
Showing 1-15 of 15
Compare
Product Number
Description
Pricing
WHA9927-047
Whatman
®
glass microfiber filters, Grade 934-AH
®
RTU,
double weight, diam. (47 mm), for solids
Expand
View Pricing
WHA9927-090
Whatman
®
glass microfiber filters, Grade 934-AH
®
RTU,
double weight, diam. (90 mm), for solids
Expand
View Pricing
WHA9907-070
Whatman
®
glass microfiber filters, Grade 934-AH
®
RTU,
diam. (70 mm), for solids
Expand
View Pricing
WHA99079436
Whatman
®
glass microfiber filters, Grade 934-AH
®
RTU,
circles, 5 decimal places, diam. (47 mm), pack of 100 ea
Expand
View Pricing
WHA3827-090
Whatman
®
glass microfiber filters, Grade 934-AH
®
RTU,
for volatiles, diam. (90 mm), pre-weighed
Expand
View Pricing
WHA4827-090
Whatman
®
glass microfiber filters, Grade 934-AH
®
RTU,
for volatiles, diam. (90 mm), pre-rinsed and ignited
Expand
View Pricing
WHA9927-070
Whatman
®
glass microfiber filters, Grade 934-AH
®
RTU,
double weight, diam. (70 mm), for solids
Expand
View Pricing
WHA3827-047
Whatman
®
glass microfiber filters, Grade 934-AH
®
RTU,
for volatiles, diam. (45 mm), pre-weighed
Expand
View Pricing
WHA4827-035
Whatman
®
glass microfiber filters, Grade 934-AH
®
RTU,
for volatiles, diam. (35 mm), pre-rinsed and ignited
Expand
View Pricing
WHA3827-035
Whatman
®
glass microfiber filters, Grade 934-AH
®
RTU,
for volatiles, diam. (35 mm), pre-weighed
Expand
View Pricing
WHA3827-042
Whatman
®
glass microfiber filters, Grade 934-AH
®
RTU,
for volatiles, diam. (42.5 mm), pre-weighed
Expand
View Pricing
WHA4827-047
Whatman
®
glass microfiber filters, Grade 934-AH
®
RTU,
for volatiles, diam. (45 mm), pre-rinsed and ignited
Expand
View Pricing
WHA4827-070
Whatman
®
glass microfiber filters, Grade 934-AH
®
RTU,
for volatiles, diam. (70 mm), pre-rinsed and ignited
Expand
View Pricing
WHA3827-070
Whatman
®
glass microfiber filters, Grade 934-AH
®
RTU,
for volatiles, diam. (70 mm), pre-weighed
Expand
View Pricing
WHA4827-042
Whatman
®
glass microfiber filters, Grade 934-AH
®
RTU,
for volatiles, diam. (42.5 mm), pre-rinsed and ignited
Expand
View Pricing