![Valine European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/347/767/33acbdb5-5aaf-4e08-8569-8350aca83998/640/33acbdb5-5aaf-4e08-8569-8350aca83998.png)
Valine
![Valine European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/347/767/33acbdb5-5aaf-4e08-8569-8350aca83998/640/33acbdb5-5aaf-4e08-8569-8350aca83998.png)
Synonym(s):
L-Valine, (S)-α-Aminoisovaleric acid, L-2-Amino-3-methylbutanoic acid
Linear Formula:
(CH3)2CHCH(NH2)CO2H
CAS Number:
Molecular Weight:
117.15
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing