Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
TSKgel
®
DEAE-5PW (13 µm) HPLC Columns
Products
Product Category
columns (2)
Manufacturer
Product Line
Separation Technique
ID (mm)
Length (cm)
Particle Size (µm)
Pore Size (Å)
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
807574
TSKgel
®
DEAE-5PW (13 µm) HPLC Columns,
L x I.D. 15 cm × 21.5 mm, DEAE (diethylaminoethyl) phase
Expand
View Pricing
814016
TSKgel
®
DEAE-5PW (13 µm) HPLC Columns,
L x I.D. 15 cm × 20 mm, DEAE (diethylaminoethyl) phase
Expand
View Pricing
814466
TSKgel
®
DEAE-5PW (13 µm) HPLC Columns,
L x I.D. 2 cm × 20 mm, DEAE (diethylaminoethyl) phase
Expand
View Pricing