
Tris(4-fluorophenyl)phosphine

Linear Formula:
(FC6H4)3P
CAS Number:
Molecular Weight:
316.26
Products
Sustainability
Feature
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing