![Trimethylphenylammonium hydroxide solution ~0.5 M (CH3)3N(OH)C6H5 in methanol, for GC derivatization, LiChropur™](/deepweb/assets/sigmaaldrich/product/structures/523/485/710a2abf-7501-40a9-9f57-aa8d8e7a64b3/640/710a2abf-7501-40a9-9f57-aa8d8e7a64b3.png)
Trimethylphenylammonium hydroxide solution
![Trimethylphenylammonium hydroxide solution ~0.5 M (CH3)3N(OH)C6H5 in methanol, for GC derivatization, LiChropur™](/deepweb/assets/sigmaaldrich/product/structures/523/485/710a2abf-7501-40a9-9f57-aa8d8e7a64b3/640/710a2abf-7501-40a9-9f57-aa8d8e7a64b3.png)
Synonym(s):
Phenyltrimethylammonium hydroxide, TMAH
Linear Formula:
(CH3)3N(OH)C6H5
CAS Number:
Molecular Weight:
153.22
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Functional Group
Reaction Type
Reagent Type
Special Grade
Technique
Showing 1-2 of 2