
Thiamethoxam

Synonym(s):
3-(2-Chloro-5-thiazolylmethyl)tetrahydro-5-methyl-N-nitro-4H-1,3,5-oxadiazin-4-imine
Empirical Formula (Hill Notation):
C8H10ClN5O3S
CAS Number:
Molecular Weight:
291.71
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2