Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
TFA-Hexylaminolinker Phosphoramidite
Synonym(s):
TFA-Hexylaminolinker Amidite
Empirical Formula (Hill Notation):
C
17
H
31
F
3
N
3
O
3
P
CAS Number:
133975-85-6
Molecular Weight:
413.42
Products
Product Category
amidites (4)
Brand
Sigma-Aldrich (4)
Formula Weight
Product Line
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
M023030
TFA-Hexylaminolinker Phosphoramidite,
configured for ABI
Expand
View Pricing
M023080
TFA-Hexylaminolinker Phosphoramidite,
configured for PerkinElmer, configured for Polygen
Expand
View Pricing
M023000
TFA-Hexylaminolinker Phosphoramidite,
Expand
View Pricing
M010800
TFA-Hexylaminolinker Phosphoramidite,
Expand
View Pricing