
Tetraallylsilane

Synonym(s):
Tetra(2-propenyl)silane
Linear Formula:
(H2C=CHCH2)4Si
CAS Number:
Molecular Weight:
192.37
Products
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing