Skip to Content
Merck
tert-Butanol SAJ first grade, ≥98.0%

tert-Butanol

tert-Butanol SAJ first grade, ≥98.0%
Synonym(s):
2-Methyl-2-propanol, tert-Butyl alcohol, Trimethyl carbinol, tert-Butanol, tert-Butanol
Linear Formula:
(CH3)3COH
CAS Number:
Molecular Weight:
74.12
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique