Skip to Content
Merck
Succinic acid European Pharmacopoeia (EP) Reference Standard

Succinic acid

Succinic acid European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Butanedioic acid, Succinic acid
Linear Formula:
HOOCCH2CH2COOH
CAS Number:
Molecular Weight:
118.09
EC Number:

Products

Product Category
Brand
Sustainability
Boiling Point (°C)
Feature
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Organoleptic Class
API Family
Reference Material Type
Special Grade
Technique
Showing 1-18 of 18
Compare
Product Number
Description
Pricing