Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Stainless Steel - 17-7PH alloy, FeCr17Ni7
Products
Manufacturer
Physical Form
Length (m)
Filter Products
Showing 1-20 of 70
Page 1 of 4
Page 1 of 4
Compare
Product Number
Description
Pricing
GF12810899
Stainless Steel - 17-7PH alloy, FeCr17Ni7,
10m wire, 0.5mm diameter
Expand
View Pricing
GF89332780
Stainless Steel - 17-7PH alloy, FeCr17Ni7,
foil, 50mm disks, 0.075mm thickness, annealed - condition a, precipitation hardenable
Expand
View Pricing
GF69319110
Stainless Steel - 17-7PH alloy, FeCr17Ni7,
foil, 100 x 100mm, 0.05mm thickness, as rolled - condition c, precipitation hardenable, light tight (lt)
Expand
View Pricing
GF61647127
Stainless Steel - 17-7PH alloy, FeCr17Ni7,
100m wire, 0.5mm diameter
Expand
View Pricing
GF74771826
Stainless Steel - 17-7PH alloy, FeCr17Ni7,
foil, 10mm disks, 0.125mm thickness, annealed - condition a, precipitation hardenable
Expand
View Pricing
GF66477291
Stainless Steel - 17-7PH alloy, FeCr17Ni7,
foil, 6mm disks, 0.075mm thickness, annealed - condition a, precipitation hardenable
Expand
View Pricing
GF11691468
Stainless Steel - 17-7PH alloy, FeCr17Ni7,
foil, 150 x 150mm, 0.05mm thickness, as rolled - condition c, precipitation hardenable, light tight (lt)
Expand
View Pricing
GF96103267
Stainless Steel - 17-7PH alloy, FeCr17Ni7,
50m wire, 0.5mm diameter
Expand
View Pricing
GF04671244
Stainless Steel - 17-7PH alloy, FeCr17Ni7,
20m wire, 0.5mm diameter
Expand
View Pricing
GF18661619
Stainless Steel - 17-7PH alloy, FeCr17Ni7,
foil, 2m coil, 14mm coil width, 0.635mm thickness, annealed - condition a, precipitation hardenable
Expand
View Pricing
GF89192481
Stainless Steel - 17-7PH alloy, FeCr17Ni7,
foil, 100 x 100mm, 0.3mm thickness, annealed - condition a, precipitation hardenable
Expand
View Pricing
GF24757713
Stainless Steel - 17-7PH alloy, FeCr17Ni7,
foil, 50 x 50mm, 0.125mm thickness, annealed - condition a, precipitation hardenable
Expand
View Pricing
GF65344619
Stainless Steel - 17-7PH alloy, FeCr17Ni7,
foil, 100 x 100mm, 0.075mm thickness, annealed - condition a, precipitation hardenable, light tight (lt)
Expand
View Pricing
GF17405782
Stainless Steel - 17-7PH alloy, FeCr17Ni7,
foil, 25 x 25mm, 0.075mm thickness, annealed - condition a, precipitation hardenable, light tight (lt)
Expand
View Pricing
GF09561898
Stainless Steel - 17-7PH alloy, FeCr17Ni7,
foil, 300 x 300mm, 0.075mm thickness, annealed - condition a, precipitation hardenable, light tight (lt)
Expand
View Pricing
GF45332546
Stainless Steel - 17-7PH alloy, FeCr17Ni7,
foil, 2m coil, 0.3mm thickness, annealed - condition a, precipitation hardenable
Expand
View Pricing
GF84155508
Stainless Steel - 17-7PH alloy, FeCr17Ni7,
foil, 2m coil, 25mm coil width, 0.15mm thickness, as rolled - condition c, precipitation hardenable
Expand
View Pricing
GF19738873
Stainless Steel - 17-7PH alloy, FeCr17Ni7,
foil, 50mm disks, 0.125mm thickness, annealed - condition a, precipitation hardenable
Expand
View Pricing
GF09757918
Stainless Steel - 17-7PH alloy, FeCr17Ni7,
foil, 25mm disks, 0.125mm thickness, annealed - condition a, precipitation hardenable
Expand
View Pricing
GF58616916
Stainless Steel - 17-7PH alloy, FeCr17Ni7,
foil, 15mm disks, 0.075mm thickness, annealed - condition a, precipitation hardenable
Expand
View Pricing
Page 1 of 4
Page 1 of 4