
Spirotetramat Metabolite BYI08330-cis-keto-hydroxy

Synonym(s):
cis-3-(2,5-Dimethylphenyl)-3-hydroxy-8-methoxy-1-azaspiro[4.5]decane-2,4-dione
Empirical Formula (Hill Notation):
C18H23NO4
CAS Number:
Molecular Weight:
317.38
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2