
Sodium propionate-13C3

Synonym(s):
Propionic acid-13C3 sodium salt
Linear Formula:
13CH313CH213CO2Na
CAS Number:
Molecular Weight:
99.04
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Isotope
Markush Class
Markush Group
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing