Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sodium peroxide
Synonym(s):
Sodium peroxide, Sodium superoxide
Empirical Formula (Hill Notation):
Na
2
O
2
CAS Number:
1313-60-6
Molecular Weight:
77.98
EC Number:
215-209-4
Products
Product Category
salts (4)
Brand
Sigma-Aldrich (3)
Supelco (1)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Quality Segment
Reagent Type
Special Grade
ACS reagent (2)
Puriss (2)
Puriss P.A. (2)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
1.06563
Sodium peroxide,
granular for analysis ACS,ISO
Expand
View Pricing
223417
Sodium peroxide,
granular, +140 mesh particle size, reagent grade, 97%
Expand
View Pricing
31445
Sodium peroxide,
puriss. p.a., ACS reagent, reag. ISO, ≥95%
Expand
View Pricing
8.14606
Sodium peroxide,
for synthesis
Expand
View Pricing
71883
Sodium peroxide,
for Wurzschmitt-decomposition, ACS reagent, beads (small), ≥95%
Expand
View Pricing