Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sodium oxalate
Synonym(s):
Ethanedioic acid sodium salt, Oxalic acid disodium salt
Linear Formula:
NaOOCCOONa
CAS Number:
62-76-0
Molecular Weight:
134.00
EC Number:
200-550-3
Products
Product Category
building blocks (2)
salts (2)
Brand
Sigma-Aldrich (4)
Supelco (1)
Formula Weight
pH Value
Physical Form
Product Line
Purity
Format
Quality Segment
Special Grade
ACS reagent (2)
Puriss (1)
Puriss P.A. (1)
Reference Material (1)
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
223433
Sodium oxalate,
ACS reagent, ≥99.5%
Expand
View Pricing
71800
Sodium oxalate,
puriss. p.a., ACS reagent, ≥99.5% (RT)
Expand
View Pricing
379735
Sodium oxalate,
≥99.99% trace metals basis
Expand
View Pricing
O0136
Sodium oxalate,
BioXtra
Expand
View Pricing
71804
Sodium oxalate,
reference material for titrimetry, certified by BAM, >99.5%
Expand
View Pricing