Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sodium diphenylamine-4-sulfonate
Synonym(s):
4-(Phenylamino)benzenesulfonic acid sodium salt, Diphenylamine-4-sulfonic acid sodium salt
Empirical Formula (Hill Notation):
C
12
H
10
NNaO
3
S
CAS Number:
6152-67-6
Molecular Weight:
271.27
EC Number:
228-165-6
Products
Formula Weight
Quality Segment
Markush Class
Markush Group
Special Grade
ACS reagent (1)
Technique
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
242969
Sodium diphenylamine-4-sulfonate,
ACS reagent
Expand
View Pricing