Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Silver nanowires
Synonym(s):
Silver nanofibers, Silver nanowhiskers, Silver nanowire
Empirical Formula (Hill Notation):
Ag
CAS Number:
7440-22-4
Molecular Weight:
107.87
EC Number:
200-661-7
Products
Brand
Sigma-Aldrich (2)
Formula Weight
Physical Form
Quality Segment
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
739421
Silver nanowires,
diam. × L 50 nm × 6 μm, 0.5% (isopropyl alcohol suspension)
Expand
View Pricing
778095
Silver nanowires,
diam. × L 200 nm × 10 μm, 0.5% (isopropyl alcohol suspension)
Expand
View Pricing
739448
Silver nanowires,
diam. × L 100 nm × 6 μm, 0.5% (isopropyl alcohol suspension)
Expand
View Pricing