Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Silver/Copper
Products
Manufacturer
Physical Form
Length (m)
Particle Size (µm)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
GF27361313
Silver/Copper,
powder, Ag 72%/Cu 28%, max. particle size 45 micron, weight 200 g
Expand
View Pricing
GF13888996
Silver/Copper,
wire, Ag 72%/Cu 28%, 1.0 mm diameter, length 2 m, temper as drawn
Expand
View Pricing
GF36678796
Silver/Copper,
wire, Ag 72%/Cu 28%, 1.0 mm diameter, length 5 m, temper as drawn
Expand
View Pricing
GF85587812
Silver/Copper,
wire, 1.0 mm diameter, Ag72%/Cu28%, temper as drawn, length 20 m
Expand
View Pricing