Skip to Content
Merck
Silicon dioxide JIS special grade

Silicon dioxide

Silicon dioxide JIS special grade
Synonym(s):
Cristobalite, Quartz, Sand, Silica
Linear Formula:
SiO2
CAS Number:
Molecular Weight:
60.08

Products

Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Length (mm)
Quality Segment
Application
Special Grade