Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Rubber hand protector
Synonym(s):
Rubber hand protector
Products
Product Category
gloves (2)
Brand
Sigma-Aldrich (1)
Manufacturer
Color
Package Size
Size
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
BAF380000002
Rubber hand protector,
green silicone, W × H 10 cm × 19 cm , 3⅞ x 7½"
Expand
View Pricing
Z104612
Rubber hand protector,
red silicone, W × W 10 cm × 4 cm , 3⅞ x 7½"
Expand
View Pricing