
(R)-(−)-3-Quinuclidinyl benzilate

Synonym(s):
(R)-(−)-QNB, (R)-(−)-Quinuclidinyl-α-hydroxydiphenylacetate
Empirical Formula (Hill Notation):
C21H23NO3
CAS Number:
Molecular Weight:
337.41
Products
Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing