Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pyrogallol
Synonym(s):
1,2,3-Trihydroxybenzene, 2,3-Dihydroxyphenol, Pyrogallic acid, Pyrogallol
Linear Formula:
C
6
H
3
(OH)
3
CAS Number:
87-66-1
Molecular Weight:
126.11
EC Number:
201-762-9
Products
Product Category
building blocks (3)
Brand
Sigma-Aldrich (5)
Supelco (1)
Boiling Point (°C)
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
analytical standard (2)
ACS reagent (1)
Puriss (1)
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
P0381
Pyrogallol,
≥98% (HPLC)
Expand
View Pricing
16040
Pyrogallol,
ACS reagent, ≥99%
Expand
View Pricing
8.22302
Pyrogallol,
for synthesis
Expand
View Pricing
254002
Pyrogallol,
ACS reagent
Expand
View Pricing
PHL89772
Pyrogallol,
phyproof
®
Reference Substance
Expand
View Pricing
24-6660
Pyrogallol,
JIS special grade, ≥99.0%
Expand
View Pricing
06931
Pyrogallol,
analytical standard
Expand
View Pricing