Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pyrex
®
VISTA
™
Griffin beakers
Synonym(s):
Corning
®
70000
Products
Product Category
beakers (10)
Manufacturer
Capacity (mL)
Package Size
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
CLS700001L
Pyrex
®
VISTA
™
Griffin beakers,
low form, capacity 1,000 mL
Expand
View Pricing
CLS70000250
Pyrex
®
VISTA
™
Griffin beakers,
low form, capacity 250 mL
Expand
View Pricing
CLS7000050
Pyrex
®
VISTA
™
Griffin beakers,
low form, capacity 50 mL
Expand
View Pricing
CLS70000100
Pyrex
®
VISTA
™
Griffin beakers,
low form, capacity 100 mL
Expand
View Pricing
CLS7000020
Pyrex
®
VISTA
™
Griffin beakers,
low form, capacity 20 mL
Expand
View Pricing
CLS7000030
Pyrex
®
VISTA
™
Griffin beakers,
low form, capacity 30 mL
Expand
View Pricing
CLS70000150
Pyrex
®
VISTA
™
Griffin beakers,
low form, capacity 150 mL
Expand
View Pricing
CLS70000600
Pyrex
®
VISTA
™
Griffin beakers,
low form, capacity 600 mL
Expand
View Pricing
CLS7000010
Pyrex
®
VISTA
™
Griffin beakers,
capacity × H × O.D. 10 mL × 33 mm × 25 mm
Expand
View Pricing
CLS70000400
Pyrex
®
VISTA
™
Griffin beakers,
low form, capacity 400 mL
Expand
View Pricing