Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pyrex
®
VISTA
™
culture tube, rimless
Synonym(s):
Corning
®
70820
Products
Product Category
tubes (11)
Color
Manufacturer
Capacity (mL)
Diameter (mm)
Cap
Package Size
Sterility
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
CLS7082016
Pyrex
®
VISTA
™
culture tube, rimless,
diam. × L 16 mm × 100 mm
Expand
View Pricing
CLS7082025
Pyrex
®
VISTA
™
culture tube, rimless,
diam. × L 25 mm × 150 mm
Expand
View Pricing
CLS7082025X
Pyrex
®
VISTA
™
culture tube, rimless,
diam. × L 25 mm × 200 mm
Expand
View Pricing
CLS7082013
Pyrex
®
VISTA
™
culture tube, rimless,
diam. × L 13 mm × 100 mm
Expand
View Pricing
CLS7082016X
Pyrex
®
VISTA
™
culture tube, rimless,
diam. × L 16 mm × 125 mm
Expand
View Pricing
CLS7082012
Pyrex
®
VISTA
™
culture tube, rimless,
diam. × L 12 mm × 75 mm
Expand
View Pricing
CLS7082020
Pyrex
®
VISTA
™
culture tube, rimless,
diam. × L 20 mm × 150 mm
Expand
View Pricing
CLS7082010
Pyrex
®
VISTA
™
culture tube, rimless,
diam. × L 10 mm × 75 mm
Expand
View Pricing
CLS7082016XX
Pyrex
®
VISTA
™
culture tube, rimless,
diam. × L 16 mm × 150 mm
Expand
View Pricing
CLS7082018
Pyrex
®
VISTA
™
culture tube, rimless,
diam. × L 18 mm × 150 mm
Expand
View Pricing
CLS7082022
Pyrex
®
VISTA
™
culture tube, rimless,
diam. × L 22 mm × 175 mm
Expand
View Pricing