Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pyrex
®
long neck culture flask
Synonym(s):
Corning
®
4442
Products
Product Category
flasks (6)
Manufacturer
Capacity (mL)
OD (mm)
Cap
Closure Type
Package Size
Sterility
Style
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
CLS4442250
Pyrex
®
long neck culture flask,
capacity 250 mL
Expand
View Pricing
CLS44421L
Pyrex
®
long neck culture flask,
capacity 1000 mL
Expand
View Pricing
CLS4442500
Pyrex
®
long neck culture flask,
capacity 500 mL
Expand
View Pricing
CLS44422L
Pyrex
®
long neck culture flask,
capacity 2000 mL
Expand
View Pricing
CLS444250
Pyrex
®
long neck culture flask,
capacity 50 mL
Expand
View Pricing
CLS4442125
Pyrex
®
long neck culture flask,
capacity 125 mL
Expand
View Pricing