Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pyrex
®
general purpose distilling apparatus, Pyrex stopper, ST joint
Synonym(s):
Corning
®
3360
Products
Product Category
flasks (2)
Manufacturer
Capacity (L)
Capacity (mL)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
CLS33601LFO
Pyrex
®
general purpose distilling apparatus, Pyrex stopper, ST joint,
stopper and flask only, capacity 1,000 mL
Expand
View Pricing
CLS3360500FO
Pyrex
®
general purpose distilling apparatus, Pyrex stopper, ST joint,
stopper and flask only, capacity 500 mL
Expand
View Pricing
CLS33601L
Pyrex
®
general purpose distilling apparatus, Pyrex stopper, ST joint,
capacity 1 L, joint: ST/NS 19/38
Expand
View Pricing