Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pyrex
®
culture tubes with screw-cap
Synonym(s):
Corning
®
9825
Products
Product Category
tubes (10)
Color
Manufacturer
OD (mm)
Package Size
Sterility
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
CLS982516X
Pyrex
®
culture tubes with screw-cap,
O.D. × L 16 mm × 125 mm
Expand
View Pricing
CLS982513
Pyrex
®
culture tubes with screw-cap,
O.D. × L 13 mm × 100 mm
Expand
View Pricing
CLS982525
Pyrex
®
culture tubes with screw-cap,
O.D. × L 25 mm × 150 mm
Expand
View Pricing
CLS982525X
Pyrex
®
culture tubes with screw-cap,
O.D. × L 25 mm × 200 mm
Expand
View Pricing
CLS982516
Pyrex
®
culture tubes with screw-cap,
O.D. × L 16 mm × 100 mm
Expand
View Pricing
CLS982538
Pyrex
®
culture tubes with screw-cap,
O.D. × L 38 mm × 200 mm
Expand
View Pricing
CLS982522
Pyrex
®
culture tubes with screw-cap,
O.D. × L 22 mm × 175 mm
Expand
View Pricing
CLS982520X
Pyrex
®
culture tubes with screw-cap,
O.D. × L 20 mm × 150 mm
Expand
View Pricing
CLS982516XX
Pyrex
®
culture tubes with screw-cap,
O.D. × L 16 mm × 150 mm
Expand
View Pricing
CLS982520
Pyrex
®
culture tubes with screw-cap,
O.D. × L 20 mm × 125 mm
Expand
View Pricing