Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
PTFE-faced silicone septa for Q-Tube
™
and QianCap
™
Products
Product Category
septa (3)
Manufacturer
Diameter (mm)
Color
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
Z567744
PTFE-faced silicone septa for Q-Tube
™
and QianCap
™
,
Septa 35 mL Low
Expand
View Pricing
Z742416
PTFE-faced silicone septa for Q-Tube
™
and QianCap
™
,
septa 12 mL, max. pressure 180 psi
Expand
View Pricing
Z567698
PTFE-faced silicone septa for Q-Tube
™
and QianCap
™
,
Septa 12 mL Low
Expand
View Pricing