
Propranolol Related Compound A

Synonym(s):
3-Naphthalen-1-yloxypropane-1,2-diol
Empirical Formula (Hill Notation):
C13H14O3
CAS Number:
Molecular Weight:
218.25
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2