
Pregabalin Related Compound A

Synonym(s):
(3R)-3-(Aminomethyl)-5-methylhexanoic acid
Empirical Formula (Hill Notation):
C8H17NO2
CAS Number:
Molecular Weight:
159.23
Products
Brand
Formula Weight
Manufacturer
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2