
Praziquantel Related Compound B

Synonym(s):
2-(Cyclohexylcarbonyl)-2,3,6,7-tetrahydro-4H-pyrazino[2,1-a]isoquinolin-4-one
Empirical Formula (Hill Notation):
C19H22N2O2
CAS Number:
Molecular Weight:
310.39
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2