Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Potassium tartrate dibasic hemihydrate
Synonym(s):
L-(+)-Tartaric acid dipotassium salt, Dipotassium tartrate hemihydrate
Linear Formula:
KOOC(CHOH)
2
COOK · 0.5H
2
O
CAS Number:
6100-19-2
Molecular Weight:
235.28
EC Number:
213-067-8
Products
Brand
Sigma-Aldrich (1)
Formula Weight
pH Value
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Optical Isomer
Special Grade
Puriss (1)
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
25510
Potassium tartrate dibasic hemihydrate,
meets analytical specification of DAC, E336, 99-102% (perchloric acid titration)
Expand
View Pricing