Skip to Content
Merck
Potassium phthalate monobasic JIS special grade, 99.8-100.2%

Potassium phthalate monobasic

Potassium phthalate monobasic JIS special grade, 99.8-100.2%
Synonym(s):
KHP, Phthalic acid monopotassium salt, Potassium biphthalate, Potassium hydrogen phthalate
Linear Formula:
HOOCC6H4COOK
CAS Number:
Molecular Weight:
204.22
EC Number:

Products

Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique