Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Poly(vinyl butyral-
co
-vinyl alcohol-
co
-vinyl acetate)
CAS Number:
27360-07-2
Products
Brand
Sigma-Aldrich (3)
Physical Form
Chemical Composition
Polymer Composition
Polymer Type
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
182567
Poly(vinyl butyral-
co
-vinyl alcohol-
co
-vinyl acetate),
average M
w
50,000-80,000 by GPC, powder
Expand
View Pricing
418420
Poly(vinyl butyral-
co
-vinyl alcohol-
co
-vinyl acetate),
average M
w
170,000-250,000, powder
Expand
View Pricing
418412
Poly(vinyl butyral-
co
-vinyl alcohol-
co
-vinyl acetate),
average M
w
90,000-120,000, powder
Expand
View Pricing