Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Polystyrene (narrow molecular weight distribution)
Synonym(s):
Polystyrene, PS
Linear Formula:
[CH
2
CH(C
6
H
5
)]
n
Products
Manufacturer
Physical Form
Quality Segment
Application
Special Grade
certified reference material (3)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
NIST1479
Polystyrene (narrow molecular weight distribution),
NIST
®
SRM
®
1479
Expand
View Pricing
NIST1478
Polystyrene (narrow molecular weight distribution),
NIST
®
SRM
®
1478
Expand
View Pricing
NIST705A
Polystyrene (narrow molecular weight distribution),
NIST
®
SRM
®
705a
Expand
View Pricing