Poly(methyl methacrylate)
Synonym(s):
PMMA, Poly(methacrylic acid methyl ester)
Linear Formula:
[CH2C(CH3)(CO2CH3)]n
CAS Number:
Products
Brand
Physical Form
Format
Quality Segment
Special Grade
Technique
Showing 1-15 of 15
Compare
Product Number
Description
Pricing