Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Platform Clamps, for Eppendorf
®
Shakers
Products
Product Category
clamps (17)
Manufacturer
Filter Products
Showing 1-17 of 17
Compare
Product Number
Description
Pricing
EPM1190-9001
Platform Clamps, for Eppendorf
®
Shakers,
for Eppendorf
®
New Brunswick Innova, I-Series and Excella shakers, for 125 mL Erlenmeyer flasks
Expand
View Pricing
EPM1190-9002
Platform Clamps, for Eppendorf
®
Shakers,
for Eppendorf
®
New Brunswick Innova, I-Series and Excella shakers, for 250 mL Erlenmeyer flasks
Expand
View Pricing
EPACFE-2800S
Platform Clamps, for Eppendorf
®
Shakers,
for Eppendorf
®
New Brunswick Innova, I-Series and Excella shakers, for 2.8 L Fernbach flasks
Expand
View Pricing
EPACE-4000S
Platform Clamps, for Eppendorf
®
Shakers,
for Eppendorf
®
New Brunswick Innova, I-Series and Excella shakers, for 4 L Erlenmeyer flasks
Expand
View Pricing
EPACE-3000S
Platform Clamps, for Eppendorf
®
Shakers,
for Eppendorf
®
New Brunswick Innova, I-Series and Excella shakers, for 3 L Erlenmeyer flasks
Expand
View Pricing
EPM1190-9003
Platform Clamps, for Eppendorf
®
Shakers,
for Eppendorf
®
New Brunswick Innova, I-Series and Excella shakers, for 500 mL Erlenmeyer flasks
Expand
View Pricing
EPACE-2000S
Platform Clamps, for Eppendorf
®
Shakers,
for Eppendorf
®
New Brunswick Innova, I-Series and Excella shakers, for 2 L Erlenmeyer flasks
Expand
View Pricing
EPM1190-9005
Platform Clamps, for Eppendorf
®
Shakers,
for Eppendorf
®
New Brunswick Innova, I-Series and Excella shakers, for 2 L Thompson flasks
Expand
View Pricing
EPACE-1000S
Platform Clamps, for Eppendorf
®
Shakers,
for Eppendorf
®
New Brunswick Innova, I-Series and Excella shakers, for 1 L Erlenmeyer flasks
Expand
View Pricing
EPM1190-9004
Platform Clamps, for Eppendorf
®
Shakers,
for Eppendorf
®
New Brunswick Innova, I-Series and Excella shakers, for 25 mL Erlenmeyer flasks
Expand
View Pricing
EPACSB-1000S
Platform Clamps, for Eppendorf
®
Shakers,
for Eppendorf
®
New Brunswick Innova 2000 shaker, for 1 L media bottles
Expand
View Pricing
EPACSB-500S
Platform Clamps, for Eppendorf
®
Shakers,
for Eppendorf
®
New Brunswick Innova 2000 shaker, for 500 mL media bottles
Expand
View Pricing
EPACE-5000S
Platform Clamps, for Eppendorf
®
Shakers,
for Excella E25/E25R, for 5 L Erlenmeyer flasks
Expand
View Pricing
EPACE-6000S
Platform Clamps, for Eppendorf
®
Shakers,
for Excella E25/E25R, for 6 L Erlenmeyer flasks
Expand
View Pricing
EPM1190-9921
Platform Clamps, for Eppendorf
®
Shakers,
for Eppendorf
®
New Brunswick Innova, I-Series and Excella shakers, starter kit
Expand
View Pricing
EPM1190-9000
Platform Clamps, for Eppendorf
®
Shakers,
for Eppendorf
®
New Brunswick Innova, I-Series and Excella shakers, for 50 mL Erlenmeyer flasks
Expand
View Pricing
EPACE-10S
Platform Clamps, for Eppendorf
®
Shakers,
for Eppendorf
®
New Brunswick Innova, I-Series and Excella shakers, for 10 mL Erlenmeyer flasks
Expand
View Pricing