
Plastic additive 11

Synonym(s):
Octadecyl 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate
Linear Formula:
[(CH3)3C]2C6H2(OH)CH2CH2CO2(CH2)17CH3
CAS Number:
Molecular Weight:
530.86
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing