
Piroctone olamine

Synonym(s):
1-Hydroxy-4-methyl-6-(2,4,4-trimethylpentyl)-2(1H)-pyridone ethanolammonium salt
Empirical Formula (Hill Notation):
C14H23NO2 · C2H7NO
CAS Number:
Molecular Weight:
298.42
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Purity
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3