
Photo-reactive Clickable cis-Sterol Probe

Empirical Formula (Hill Notation):
C30H46N2O3
Molecular Weight:
482.70
Products
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing