
Phosphoric acid

Products
Product Category
Brand
Sustainability
Boiling Point (°C)
Feature
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Technique
Showing 1-12 of 12
Compare
Product Number
Description
Pricing