Phenytoin Related Compound B
Synonym(s):
α-[(Aminocarbonyl)amino]-α-phenylbenzeneacetic acid
Empirical Formula (Hill Notation):
C15H14N2O3
CAS Number:
Molecular Weight:
270.28
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2