
Phenobarbital

Synonym(s):
5-Ethyl-5-phenyl-2,4,6-pyrimidinetrione, 5-Ethyl-5-phenylbarbituric acid, Luminal
Empirical Formula (Hill Notation):
C12H12N2O3
CAS Number:
Molecular Weight:
232.24
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade