![Pepstatin from Streptomyces species](/deepweb/assets/sigmaaldrich/product/structures/253/546/41bdeacc-9913-4ac3-96f3-726c5c85710a/640/41bdeacc-9913-4ac3-96f3-726c5c85710a.png)
Pepstatin
![Pepstatin from Streptomyces species](/deepweb/assets/sigmaaldrich/product/structures/253/546/41bdeacc-9913-4ac3-96f3-726c5c85710a/640/41bdeacc-9913-4ac3-96f3-726c5c85710a.png)
Synonym(s):
Pepstatin A
Empirical Formula (Hill Notation):
C34H63N5O9
CAS Number:
Molecular Weight:
685.89
Products
Biological Source
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing