Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
PCR strip tube caps
Products
Product Category
caps (3)
Color
Manufacturer
Sterility
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
AXYPCR2CPRTC
PCR strip tube caps,
for real time PCR, thin wall, size 0.2 mL, pkg of 10x125strips/cs, clear
Expand
View Pricing
AXYPCR02CPA
PCR strip tube caps,
thin wall, 8 per strip, size 0.2 mL, assorted colors, pkg of 10x125strips/cs
Expand
View Pricing
AXYPCR02FCPC
PCR strip tube caps,
flat with writing surface, thin wall, size 0.2 mL, clear, pkg of 10x125strips/cs
Expand
View Pricing