Skip to Content
Merck
Patent blue V sodium salt suitable for microscopy

Patent blue V sodium salt

Patent blue V sodium salt suitable for microscopy
Synonym(s):
Acid blue 3 sodium salt, Food Blue 5 sodium salt
Empirical Formula (Hill Notation):
C27H31N2NaO7S2
CAS Number:
Molecular Weight:
582.66
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Application
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing