
Paraldehyde

Synonym(s):
2,4,6-Trimethyl-1,3,5-trioxacyclohexane, s-Trimethyltrioxymethylene, Acetaldehyde trimer, NSC 9799, Paracetaldehyde, Paraldehyde
Empirical Formula (Hill Notation):
C6H12O3
CAS Number:
Molecular Weight:
132.16
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing