ortho-Phosphoric acid
Synonym(s):
Phosphoric acid, Orthophosphoric acid
Linear Formula:
H3PO4
CAS Number:
Molecular Weight:
98.00
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Agency
Quality Segment
Application
Markush Class
Markush Group
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing