Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ohaus
®
pH Meter
Products
Manufacturer
Package Size
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
OH30057496
Ohaus
®
pH Meter,
Model ST2100-F, AC/DC input 120 V, US 3-pin plug
Expand
View Pricing
OH30058489
Ohaus
®
pH Meter,
Model ST3100-F, AC/DC input 120 V, US 3-pin plug
Expand
View Pricing
OH83033961
Ohaus
®
pH Meter,
Model ST300
Expand
View Pricing
OH30129895
Ohaus
®
pH Meter,
Model ST5000-B, AC/DC input 120 V, US 3-pin plug
Expand
View Pricing
OH30129896
Ohaus
®
pH Meter,
Model ST5000-F, AC/DC input 120 V, US 3-pin plug
Expand
View Pricing
OH30219114
Ohaus
®
pH Meter,
Model ST300-G
Expand
View Pricing
OH83033962
Ohaus
®
pH Meter,
Model ST300-B
Expand
View Pricing